CAS RN 76801-85-9
CAS RN 76801-85-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
6-Demethylazithromycin (Azaerythromycin A), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Azathramycin, MedChemExpress
MedChemExpress Azathramycin (Azaerythromycin A) is an antibiotic and targets ribosome.