CAS RN 770691-21-9
CAS RN 770691-21-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gadobutrol, MedChemExpress
MedChemExpress Gadobutrol (Gd-DO3A-butrol; ZK 135079) is a nonionic paramagnetic macrocyclic gadolinium-based contrast agent that can be used for magnetic resonance imaging (MRI).