accessibility menu, dialog, popup

UserName

CAS RN 771573-01-4

CH3NH2F

CAS RN 771573-01-4

IUPAC Name: 1-(2-fluoro-4-methylphenyl)methanamine
Synonyms: 1-(2-fluoro-4-methylphenyl)methanamine
Molecular Weight (g/mol): 139.17
Molecular Formula: C8H10FN
InChi Key: QRRUYSDAUXURDL-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results