accessibility menu, dialog, popup

UserName

FFFSClOO

CAS RN 776-04-5

IUPAC Name: 2-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 2-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 244.61
Molecular Formula: C7H4ClF3O2S
InChi Key: ZIZGWNOAHUCACM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1S(Cl)(=O)=O
Boiling Point 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results