accessibility menu, dialog, popup

UserName

CAS RN 7770-78-7

H3CO(R)(R)OOOCH3OH3CHO

CAS RN 7770-78-7

IUPAC Name: (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(4-hydroxy-3-methoxyphenyl)methyl]oxolan-2-one
Synonyms: (-)-arctigenin arctigenin
Molecular Weight (g/mol): 372.42
Molecular Formula: C21H24O6
InChi Key: NQWVSMVXKMHKTF-JKSUJKDBSA-N
SMILES: COC1=CC=C(C[C@H]2COC(=O)[C@@H]2CC2=CC=C(O)C(OC)=C2)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)