accessibility menu, dialog, popup

UserName

H3COONNOOCH3H3C

CAS RN 77732-09-3

IUPAC Name: N-(2,6-dimethylphenyl)-2-methoxy-N-(2-oxo-1,3-oxazolidin-3-yl)acetamide
Synonyms: oxadixyl
Molecular Weight (g/mol): 278.31
Molecular Formula: C14H18N2O4
InChi Key: UWVQIROCRJWDKL-UHFFFAOYSA-N
SMILES: COCC(=O)N(N1CCOC1=O)C1=C(C)C=CC=C1C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Concentration 
  • (4)
Solvent or Matrix 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)