accessibility menu, dialog, popup

UserName

H3COONH

CAS RN 778-82-5

IUPAC Name: ethyl 2-(1H-indol-3-yl)acetate
Synonyms: ethyl 2-(1H-indol-3-yl)acetate
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: HUDBDWIQSIGUDI-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (5)
  • (1)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (3)
  • (5)
  • (9)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)