accessibility menu, dialog, popup

UserName

CAS RN 7785-70-8

CH3(R)(R)H3CH3C

CAS RN 7785-70-8

IUPAC Name: (1R,5R)-2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Synonyms: (+)-α-pinene (1R,5R)-α-pinene α-pinene(dextro) D-α-pinene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: GRWFGVWFFZKLTI-RKDXNWHRSA-N
SMILES: CC1=CC[C@@H]2C[C@H]1C2(C)C
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)