CAS RN 7786-34-7
CAS RN 7786-34-7
IUPAC Name:
methyl (2E)-3-[(dimethoxyphosphoryl)oxy]but-2-enoate
Synonyms:
mevinphos (trans)
Molecular Weight (g/mol):
224.15
Molecular Formula:
C7H13O6P
InChi Key:
GEPDYQSQVLXLEU-AATRIKPKSA-N
SMILES:
COC(=O)\C=C(/C)OP(=O)(OC)OC
1
–
4
of
4
results
2
Mevinphos, TRC
CAS: 7786-34-7 Molecular Formula: C7 H13 O6 P Molecular Weight (g/mol): 224.15 Synonym: Crotonic acid, 3-hydroxy-, methyl ester, dimethyl phosphate (6CI,8CI),1-Methoxycarbonyl-1-propen-2-yl dimethyl phosphate,2-Carbomethoxy-1-methylvinyl dimethyl phosphate,2-Methoxycarbonyl-1-methylvinyl dimethyl phosphate,CMDP,Fosdrin,Gesfid,Meniphos,Mevinphos,NSC 46470,O,O-Dimethyl O-(1-carbomethoxy-1-propen-2-yl) phosphate,PD 5,PD 5 (pesticide),Phosdrin,Phosdrin W 10 IUPAC Name: methyl 3-dimethoxyphosphoryloxybut-2-enoate SMILES: COC(=O)C=C(C)OP(=O)(OC)OC