CAS RN 779331-47-4
IUPAC Name:
[2-(benzyloxy)-5-fluorophenyl]boronic acid
Synonyms:
2-(benzyloxy)-5-fluorophenylboronic acid
Molecular Weight (g/mol):
246.04
Molecular Formula:
C13H12BFO3
InChi Key:
ULIJNMCAPASXHA-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(F)=CC=C1OCC1=CC=CC=C1
1
–
1
of
1
results
1
2-Benzyloxy-5-fluorobenzeneboronic acid, 98%, Thermo Scientific™
CAS: 779331-47-4 Molecular Formula: C13H12BFO3 Molecular Weight (g/mol): 246.044 MDL Number: MFCD03788403 InChI Key: ULIJNMCAPASXHA-UHFFFAOYSA-N Synonym: 2-benzyloxy-5-fluorophenylboronic acid,2-benzyloxy-5-fluorobenzeneboronic acid,2-benzyloxy-5-fluorophenyl boronic acid,2-benzyloxy-5-fluorophenylboronic acid contains varying amounts of anhydride,2-benzyloxy-5-fluorophenylboronicacid,acmc-209pct,2-benzyloxy-5-fluoro-phenyl boronic acid,5-fluoro-2-phenylmethoxyphenyl boronic acid PubChem CID: 2782666 IUPAC Name: (5-fluoro-2-phenylmethoxyphenyl)boronic acid SMILES: B(C1=C(C=CC(=C1)F)OCC2=CC=CC=C2)(O)O