accessibility menu, dialog, popup

UserName

H3CH3CCH3OONSiH3CCH3OH

CAS RN 784161-48-4

IUPAC Name: tert-butyl 2-(hydroxydimethylsilyl)-1H-indole-1-carboxylate
Synonyms: tert-butyl 2-(hydroxydimethylsilyl)indole-1-carboxylate
Molecular Weight (g/mol): 291.42
Molecular Formula: C15H21NO3Si
InChi Key: ARJIKJGACIMUOM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C2=CC=CC=C2C=C1[Si](C)(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results