accessibility menu, dialog, popup

UserName

CAS RN 78619-84-8

IUPAC Name: hydrogen 4,4'-bipiperidine chloride
Synonyms: hydrogen 4,4'-bipiperidine chloride
Molecular Weight (g/mol): 204.74
Molecular Formula: C10H21ClN2
InChi Key: YZYKKCNABMKECM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1CC(CCN1)C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results