accessibility menu, dialog, popup

UserName

CAS RN 792-74-5

IUPAC Name: 4,4'-dimethyl [1,1'-biphenyl]-4,4'-dicarboxylate
Synonyms: 4,4'-dimethyl [1,1'-biphenyl]-4,4'-dicarboxylate
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: BKRIRZXWWALTPU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results