accessibility menu, dialog, popup

UserName

CAS RN 79241-46-6

H3COO(R)CH3OONFFF

CAS RN 79241-46-6

IUPAC Name: butyl (2R)-2-(4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}phenoxy)propanoate
Synonyms: fluazifop-P-butyl (iso)
Molecular Weight (g/mol): 383.37
Molecular Formula: C19H20F3NO4
InChi Key: VAIZTNZGPYBOGF-CYBMUJFWSA-N
SMILES: CCCCOC(=O)[C@@H](C)OC1=CC=C(OC2=CC=C(C=N2)C(F)(F)F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results