accessibility menu, dialog, popup

UserName

CAS RN 79271-56-0

CH3SiCH3H3COSOOFFF

CAS RN 79271-56-0

IUPAC Name: triethylsilyl trifluoromethanesulfonate
Synonyms: triethylsilyl trifluoromethanesulfonate
Molecular Weight (g/mol): 264.33
Molecular Formula: C7H15F3O3SSi
InChi Key: STMPXDBGVJZCEX-UHFFFAOYSA-N
SMILES: CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)