CAS RN 79455-30-4
CAS RN 79455-30-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nicaraven, MedChemExpress
MedChemExpress Nicaraven is a novel chemically synthesized hydroxyl radical-specific scavenger.