accessibility menu, dialog, popup

UserName

CAS RN 79544-27-7

BrNF

CAS RN 79544-27-7

IUPAC Name: 2-bromo-6-fluorobenzonitrile
Synonyms: 2-bromo-6-fluorobenzonitrile
Molecular Weight (g/mol): 200.01
Molecular Formula: C7H3BrFN
InChi Key: IELGUZKHALDFOO-UHFFFAOYSA-N
SMILES: FC1=C(C#N)C(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results