CAS RN 79646-00-7
CAS RN 79646-00-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Dechloro Sertraline Hydrochloride, TRC
CAS: 79646-00-7 Molecular Formula: C17H19Cl2N Molecular Weight (g/mol): 308.25 Synonym: (1S-cis)-4-(4-Chlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine Hydrochloride; IUPAC Name: (1S,4S)-4-(4-chlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride SMILES: CN[C@H]1CC[C@H](C2=CC=CC=C12)C3=CC=C(C=C3)Cl.Cl