accessibility menu, dialog, popup

UserName

H3COOHNHCl

CAS RN 79854-42-5

IUPAC Name: ethyl 2,3-dihydro-1H-indole-2-carboxylate hydrochloride
Synonyms: ethyl 2,3-dihydro-1H-indole-2-carboxylate hydrochloride
Molecular Weight (g/mol): 227.69
Molecular Formula: C11H14ClNO2
InChi Key: VTNFLVJEOPKVIC-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C1CC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results