accessibility menu, dialog, popup

UserName

CAS RN 80-40-0

CH3OSOOCH3

CAS RN 80-40-0

IUPAC Name: ethyl 4-methylbenzene-1-sulfonate
Synonyms: ethyl-P-methylbenzenesulfonate
Molecular Weight (g/mol): 200.25
Molecular Formula: C9H12O3S
InChi Key: VRZVPALEJCLXPR-UHFFFAOYSA-N
SMILES: CCOS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)