accessibility menu, dialog, popup

UserName

CAS RN 8004-87-3

H3CNHCNCH3H3CNCH3CH3

CAS RN 8004-87-3

IUPAC Name: bis[4-(dimethylamino)phenyl][4-(methylamino)phenyl]methylium
Synonyms: colouring agent CI 42535 (basic violet 1)
Molecular Weight (g/mol): 358.51
Molecular Formula: C24H28N3
InChi Key: HJPVVBCICYOONZ-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Color 
  • (6)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (5)