CAS RN 80054-40-6
CAS RN 80054-40-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(E,E)-2,6-Dimethyl-2,6-octadienedial, TRC
CAS: 80054-40-6 Molecular Formula: C10H14O2 Molecular Weight (g/mol): 166.22 Synonym: (2E,6E)-2,6-Dimethyl-2,6-octadienedial IUPAC Name: (2E,6E)-2,6-dimethylocta-2,6-dienedial SMILES: O=C/C(C)=C/CC/C(C)=C/C=O