accessibility menu, dialog, popup

UserName

CAS RN 80143-71-1

IUPAC Name: 4-fluoro-N-[(4-methoxyphenyl)methyl]aniline
Synonyms: 4-fluoro-N-[(4-methoxyphenyl)methyl]aniline
Molecular Weight (g/mol): 231.27
Molecular Formula: C14H14FNO
InChi Key: GSIXDRVRPADHPJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(CNC2=CC=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results