accessibility menu, dialog, popup

UserName

OOSOO

CAS RN 81-08-3

IUPAC Name: 3H-2,1λ⁶-benzoxathiole-1,1,3-trione
Synonyms: 2,1λ⁶-benzoxathiole-1,1,3-trione
Molecular Weight (g/mol): 184.17
Molecular Formula: C7H4O4S
InChi Key: NCYNKWQXFADUOZ-UHFFFAOYSA-N
SMILES: O=C1OS(=O)(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)