accessibility menu, dialog, popup

UserName

OHOOHONOONOO

CAS RN 81-55-0

IUPAC Name: 1,8-dihydroxy-4,5-dinitro-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,8-dihydroxy-4,5-dinitroanthracene-9,10-dione
Molecular Weight (g/mol): 330.21
Molecular Formula: C14H6N2O8
InChi Key: GJCHQJDEYFYWER-UHFFFAOYSA-N
SMILES: OC1=CC=C(C2=C1C(=O)C1=C(O)C=CC(=C1C2=O)[N+]([O-])=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)