accessibility menu, dialog, popup

UserName

BrHNH2NHCl

CAS RN 81868-12-4

IUPAC Name: 2-(5-bromo-1H-indol-3-yl)ethan-1-amine hydrochloride
Synonyms: 2-(5-bromo-1H-indol-3-yl)ethanamine hydrochloride
Molecular Weight (g/mol): 275.57
Molecular Formula: C10H12BrClN2
InChi Key: QUPXCOYSKSUJPX-UHFFFAOYSA-N
SMILES: Cl.NCCC1=CNC2=CC=C(Br)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results