accessibility menu, dialog, popup

UserName

CAS RN 82097-50-5

CH3ONCH3NHNOHNSOOOClN

CAS RN 82097-50-5

IUPAC Name: 1-[2-(2-chloroethoxy)benzenesulfonyl]-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea
Synonyms: triasulfuron
Molecular Weight (g/mol): 401.82
Molecular Formula: C14H16ClN5O5S
InChi Key: XOPFESVZMSQIKC-UHFFFAOYSA-N
SMILES: COC1=NC(C)=NC(NC(=O)NS(=O)(=O)C2=CC=CC=C2OCCCl)=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Concentration 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results