accessibility menu, dialog, popup

UserName

FNClCl

CAS RN 823-56-3

IUPAC Name: 3,5-dichloro-2-fluoropyridine
Synonyms: 3,5-dichloro-2-fluoropyridine
Molecular Weight (g/mol): 165.98
Molecular Formula: C5H2Cl2FN
InChi Key: FHQWUIZMJXPGRG-UHFFFAOYSA-N
SMILES: FC1=NC=C(Cl)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results