accessibility menu, dialog, popup

UserName

CAS RN 824-72-6

ClPClO

CAS RN 824-72-6

IUPAC Name: phenylphosphonoyl dichloride
Synonyms: phosphonic dichloride, phenyl-
Molecular Weight (g/mol): 194.98
Molecular Formula: C6H5Cl2OP
InChi Key: IBDMRHDXAQZJAP-UHFFFAOYSA-N
SMILES: ClP(Cl)(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)