CAS RN 82414-35-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N1,N8-Diacetylspermidine, TRC
CAS: 82414-35-5 Molecular Formula: C11H23N3O2 Molecular Weight (g/mol): 229.32 Synonym: N-[3-[[4-(Acetylamino)butyl]amino]propyl]acetamide,N1,8-Diacetylspermidine SMILES: O=C(C)NCCCCNCCCNC(C)=O