CAS RN 82417-45-6
IUPAC Name:
2,3-dichlorobenzene-1-sulfonyl chloride
Synonyms:
2,3-dichlorobenzenesulfonyl chloride
Molecular Weight (g/mol):
245.5
Molecular Formula:
C6H3Cl3O2S
InChi Key:
PQODWTNHDKDHIW-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(=C1Cl)S(Cl)(=O)=O
1
–
2
of
2
results
1
2,3-Dichlorobenzenesulfonyl chloride, 98%
CAS: 82417-45-6 Molecular Formula: C6H3Cl3O2S Molecular Weight (g/mol): 245.498 MDL Number: MFCD00051844 InChI Key: PQODWTNHDKDHIW-UHFFFAOYSA-N Synonym: 2,3-dichlorobenzene-1-sulfonyl chloride,2,3-dichlorobenzenesulphonyl chloride,2,3-dichloro-benzenesulfonyl chloride,2,3-dichlorobenzenesulphonylchloride,2,3-dichlorophenylsulfonyl chloride,benzenesulfonyl chloride, 2,3-dichloro-9ci,2,3-dichlorobenzenesulfonylchloride,pubchem5114,buttpark 9357-48,acmc-1bl9r PubChem CID: 2734222 IUPAC Name: 2,3-dichlorobenzenesulfonyl chloride SMILES: C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl