accessibility menu, dialog, popup

UserName

HOHOO

CAS RN 828-01-3

IUPAC Name: 2-hydroxy-3-phenylpropanoic acid
Synonyms: β-phenyllactic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: VOXXWSYKYCBWHO-UHFFFAOYNA-N
SMILES: OC(CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
  • (2)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (4)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)