accessibility menu, dialog, popup

UserName

CAS RN 83-56-7

OHOH

CAS RN 83-56-7

IUPAC Name: naphthalene-1,5-diol
Synonyms: 1,5-naphthalenediol
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: BOKGTLAJQHTOKE-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C(O)C=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)