accessibility menu, dialog, popup

UserName

CAS RN 830-79-5

H3COOCH3OOH3C

CAS RN 830-79-5

IUPAC Name: 2,4,6-trimethoxybenzaldehyde
Synonyms: 2,4,6-trimethoxybenzaldehyde
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: CRBZVDLXAIFERF-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C=O)C(OC)=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)