accessibility menu, dialog, popup

UserName

CAS RN 834914-39-5

CH3OOOH

CAS RN 834914-39-5

IUPAC Name: ethyl 1-(hydroxymethyl)cyclohexane-1-carboxylate
Synonyms: ethyl 1-(hydroxymethyl)cyclohexane-1-carboxylate
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: CHMBRLRRLJHXRK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1(CO)CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results