accessibility menu, dialog, popup

UserName

CH3CH3H3CSOOO

CAS RN 835617-36-2

IUPAC Name: 2,4,6-trimethylbenzene-1-sulfonate
Synonyms: 2,4,6-trimethylbenzenesulfonate
Molecular Weight (g/mol): 199.24
Molecular Formula: C9H11O3S
InChi Key: LXFQSRIDYRFTJW-UHFFFAOYSA-M
SMILES: CC1=CC(C)=C(C(C)=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results