accessibility menu, dialog, popup

UserName

H3CON(E/Z)

CAS RN 836-41-9

IUPAC Name: 1-(4-methoxyphenyl)-N-phenylmethanimine
Synonyms: 1-(4-methoxyphenyl)-N-phenylmethanimine
Molecular Weight (g/mol): 211.26
Molecular Formula: C14H13NO
InChi Key: MSWPGMRTURVKRJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=NC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results