CAS RN 83741-76-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Pentanoic-5,5,5-d3 Acid, TRC
CAS: 83741-76-8 Molecular Formula: C5 D3 H7 O2 Molecular Weight (g/mol): 105.15 Synonym: Pentanoic-5,5,5-d3 acid (9CI),δ-Trideuteropentanoic acid,Pentanoic acid-5,5,5-d3,Valeric acid-d3,Valeric-δ,δ,δ-d3 acid,Pentanoic-5,5,5-D3 acid IUPAC Name: 5,5,5-trideuteriopentanoic acid SMILES: [2H]C([2H])([2H])CCCC(=O)O
2
Pentanoic-5,5,5-d3 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.