CAS RN 838-07-3
CAS RN 838-07-3
IUPAC Name:
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2-dihydropyrimidin-2-one
Synonyms:
5-methyldeoxycytidine
Molecular Weight (g/mol):
241.25
Molecular Formula:
C10H15N3O4
InChi Key:
LUCHPKXVUGJYGU-XLPZGREQSA-N
SMILES:
CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N=C1N
1
–
2
of
2
results
2
5-Methyl-2'-deoxycytidine, MP Biomedicals™
CAS: 838-07-3 MDL Number: MFCD00006549 InChI Key: LUCHPKXVUGJYGU-XLPZGREQSA-N PubChem CID: 440055 ChEBI: CHEBI:47876 IUPAC Name: 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one SMILES: CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O