accessibility menu, dialog, popup

UserName

CAS RN 84025-81-0

CH3OHN

CAS RN 84025-81-0

IUPAC Name: 2-(methoxymethyl)pyrrolidine
Synonyms: 2-(methoxymethyl)pyrrolidine
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: CHPRFKYDQRKRRK-UHFFFAOYNA-N
SMILES: COCC1CCCN1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)