accessibility menu, dialog, popup

UserName

CAS RN 84087-01-4

HOOClClN

CAS RN 84087-01-4

IUPAC Name: 3,7-dichloroquinoline-8-carboxylic acid
Synonyms: quinclorac
Molecular Weight (g/mol): 242.06
Molecular Formula: C10H5Cl2NO2
InChi Key: FFSSWMQPCJRCRV-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2N=CC(Cl)=CC2=CC=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results