CAS RN 84228-93-3
IUPAC Name:
3-(pyridin-4-yl)prop-2-enoic acid
Synonyms:
3-(pyridin-4-yl)prop-2-enoic acid
Molecular Weight (g/mol):
149.15
Molecular Formula:
C8H7NO2
InChi Key:
SSAYTINUCCRGDR-UHFFFAOYSA-N
SMILES:
OC(=O)C=CC1=CC=NC=C1
1
–
1
of
1
results
1
trans-3-(4-Pyridyl)acrylic acid, 97%
CAS: 84228-93-3 Molecular Formula: C8H7NO2 Molecular Weight (g/mol): 149.149 MDL Number: MFCD00040746 InChI Key: SSAYTINUCCRGDR-OWOJBTEDSA-N Synonym: 3-4-pyridyl acrylic acid,trans-3-4-pyridyl acrylic acid,3-pyridin-4-yl-acrylic acid,4-pyridineacrylic acid,e-3-pyridin-4-yl acrylic acid,3-pyridin-4-yl acrylic acid,2e-3-pyridin-4-ylacrylic acid,3-4-pyridine acrylic acid,2e-3-pyridin-4-yl prop-2-enoic acid,e-3-pyridin-4-yl-acrylic acid PubChem CID: 736782 IUPAC Name: (E)-3-pyridin-4-ylprop-2-enoic acid SMILES: C1=CN=CC=C1C=CC(=O)O