accessibility menu, dialog, popup

UserName

CAS RN 847560-49-0

CH3OBHOOH

CAS RN 847560-49-0

IUPAC Name: [4-(benzyloxy)-2-methylphenyl]boronic acid
Synonyms: 4-(benzyloxy)-2-methylphenylboronic acid
Molecular Weight (g/mol): 242.08
Molecular Formula: C14H15BO3
InChi Key: VCDFDGOVNBMYRW-UHFFFAOYSA-N
SMILES: CC1=CC(OCC2=CC=CC=C2)=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results