accessibility menu, dialog, popup

UserName

CH3NNH3CCH3OPOOOH3C

CAS RN 848641-69-0

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium diethyl phosphate
Synonyms: 1-ethyl-3-methylimidazolium diethyl phosphate
Molecular Weight (g/mol): 264.26
Molecular Formula: C10H21N2O4P
InChi Key: HQWOEDCLDNFWEV-UHFFFAOYSA-M
SMILES: CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results