accessibility menu, dialog, popup

UserName

CAS RN 849773-97-3

A(a)

CAS RN 849773-97-3

IUPAC Name: λ²-iron(2+) bis(1-[2,5-bis(propan-2-yl)phospholan-1-yl]cyclopenta-2,4-dien-1-ide) cycloocta-1,5-diene tetrafluoroboranuide rhodium
Synonyms: λ²-iron(2+) 1,5-cyclooctadiene bis(1-(2,5-diisopropylphospholan-1-yl)cyclopenta-2,4-dien-1-ide) rhodium tetrafluoroborate
Molecular Weight (g/mol): 824.4
Molecular Formula: C38H60BF4FeP2Rh
InChi Key: MFSWTFGIDMKBOY-UHFFFAOYNA-N
SMILES: [Fe++].[Rh].F[B-](F)(F)F.C1CC=CCCC=C1.CC(C)C1CCC(C(C)C)P1[C-]1C=CC=C1.CC(C)C1CCC(C(C)C)P1[C-]1C=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results