accessibility menu, dialog, popup

UserName

HOOOClNOO

CAS RN 85-54-1

IUPAC Name: 2-(4-chloro-3-nitrobenzoyl)benzoic acid
Synonyms: 2-(4-chloro-3-nitrobenzoyl)benzoic acid
Molecular Weight (g/mol): 305.67
Molecular Formula: C14H8ClNO5
InChi Key: RITAQDHCJBLSSL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results