CAS RN 85-79-0
CAS RN 85-79-0
IUPAC Name:
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide
Synonyms:
dibucaine nupercainal cinchocaine nupercaine dibucain dermacaine heavy solution nupercaine sovcaine dibucaine base cinchocainum
Molecular Weight (g/mol):
343.47
Molecular Formula:
C20H29N3O2
InChi Key:
PUFQVTATUTYEAL-UHFFFAOYSA-N
SMILES:
CCCCOC1=CC(C(=O)NCCN(CC)CC)=C2C=CC=CC2=N1
1
–
1
of
1
results
1
Cinchocaine, TRC
CAS: 85-79-0 Molecular Formula: C20 H29 N3 O2 Molecular Weight (g/mol): 343.46 Synonym: 4-Quinolinecarboxamide, 2-butoxy-N-[2-(diethylamino)ethyl]-,Cinchoninamide, 2-butoxy-N-[2-(diethylamino)ethyl]- (8CI),2-Butoxy-N-[2-(diethylamino)ethyl]-4-quinolinecarboxamide,2-Butoxy-N-[2-(diethylamino)ethyl]cinchoninamide,2-Butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide,Cinchocaine,Dermacaine,Dibucaine,Dibucaine base,N-[2-(Diethylamino)ethyl]-2-butoxycinchoninamide,NSC 159055,Nupercainal,Nupercaine,Percamine IUPAC Name: 2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide SMILES: CCCCOc1cc(C(=O)NCCN(CC)CC)c2ccccc2n1