accessibility menu, dialog, popup

UserName

CAS RN 850375-15-4

CH3NNNH2CH3

CAS RN 850375-15-4

IUPAC Name: 1-(1,4-dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl)methanamine
Synonyms: 1-(1,4-dimethyl-2,3-dihydroquinoxalin-6-yl)methanamine
Molecular Weight (g/mol): 191.28
Molecular Formula: C11H17N3
InChi Key: TWGYATHIWDUKGY-UHFFFAOYSA-N
SMILES: CN1CCN(C)C2=C1C=CC(CN)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results