accessibility menu, dialog, popup

UserName

CAS RN 850589-57-0

BFOCH3CH3HOOH

CAS RN 850589-57-0

IUPAC Name: [3-fluoro-5-(2-methylpropoxy)phenyl]boronic acid
Synonyms: 3-fluoro-5-(2-methylpropoxy)phenylboronic acid
Molecular Weight (g/mol): 212.03
Molecular Formula: C10H14BFO3
InChi Key: KLKWZMKGTIQLOG-UHFFFAOYSA-N
SMILES: CC(C)COC1=CC(F)=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results