accessibility menu, dialog, popup

UserName

FFO

CAS RN 85118-07-6

IUPAC Name: (3,4-difluorophenyl)(phenyl)methanone
Synonyms: (3,4-difluorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 218.2
Molecular Formula: C13H8F2O
InChi Key: ZJTYHSBOZAQQGF-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1F)C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results